Proteintech
Manufacturer Code:550051AP150UL
Catalog # 50-173-5926
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Histone-lysine N-methyltransferase SETD1B; hSET1B; hSET1B KIAA1076 KMT2G Lysine N methyltransferase 2G SET domain containing 1B Set1B SETD1B; Lysine N-methyltransferase 2G; SET domain-containing protein 1B
Gene Aliases: AA516740; BC035291; KIAA1076; KMT2G; mKIAA1076; SET1B; SETD1B
UniProt ID: (Human) Q9UPS6, (Mouse) Q8CFT2
Entrez Gene ID: (Human) 23067, (Mouse) 208043
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.