Novus Biologicals
Manufacturer Code:NBP274144C
Catalog # NBP274144C
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7B7 |
Immunogen | Recombinant protein fragment corresponding to amino acids 1-320 of human SETD1A (NP_055527) produced in E.coli. |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Histone-lysine N-methyltransferase SETD1A; histone-lysine N-methyltransferase SETD1A hSET1A KIAA0339EC 2.1.1.43 KMT2FSET1 Lysine N-methyltransferase 2F SET domain containing 1A SET domain-containing protein 1A Set1 Set1/Ash2 histone methyltransferase complex subunit SET1 SET1A; hSET1A; Lysine N-methyltransferase 2F; SET domain-containing protein 1A; Set1/Ash2 histone methyltransferase complex subunit SET1
Gene Aliases: KIAA0339; KMT2F; SET1; SET1A; SETD1A
UniProt ID: (Human) O15047
Entrez Gene ID: (Human) 9739
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.