Novus Biologicals
Manufacturer Code:NBP276401
Catalog # NBP276401
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | BLR024E |
Immunogen | Immunogen sequence between amino acids 1200 and 1250 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 2-8°C/ 1 year from date of receipt |
Rabbit Monoclonal Antibody
Protein Aliases: Histone-lysine N-methyltransferase SETD1A; histone-lysine N-methyltransferase SETD1A hSET1A KIAA0339EC 2.1.1.43 KMT2FSET1 Lysine N-methyltransferase 2F SET domain containing 1A SET domain-containing protein 1A Set1 Set1/Ash2 histone methyltransferase complex subunit SET1 SET1A; hSET1A; Lysine N-methyltransferase 2F; SET domain-containing protein 1A; Set1/Ash2 histone methyltransferase complex subunit SET1
Gene Aliases: KIAA0339; KMT2F; SET1; SET1A; SETD1A
UniProt ID: (Human) O15047
Entrez Gene ID: (Human) 9739
Molecular Function: DNA binding protein methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.