Invitrogen
Manufacturer Code:PA5116810
Catalog # PIPA5116810
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human SET1A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BC010250 Histone-lysine N-methyltransferase SETD1A hSET1A KIAA0339 KMT2F Lysine N-methyltransferase 2F mKIAA0339 mNSC1 non-selective cation channel 1 Nsccn1 SET domain containing 1A SET domain-containing protein 1A SET1 Set1/Ash2 histone methyltransferase complex subunit SET1 SET1A SETD1A; Histone-lysine N-methyltransferase SETD1A; hSET1A; Lysine N-methyltransferase 2F; non-selective cation channel 1; SET domain-containing protein 1A; Set1/Ash2 histone methyltransferase complex subunit SET1
Gene Aliases: BC010250; KIAA0339; KMT2F; mKIAA0339; mNSC1; Nsccn1; SET1; SET1A; SETD1A
UniProt ID: (Human) O15047, (Mouse) E9PYH6
Entrez Gene ID: (Human) 9739, (Mouse) 233904
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.