Abnova
Manufacturer Code:PAB14083
Catalog # 89-116-919
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A mixture of synthetic peptides (conjugated with KLH),of which two correspond to N-terminus and one to internal region of mouse Setd1a. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Setd1a.
For Research Use Only
Protein Aliases: BC010250; Histone-lysine N-methyltransferase SETD1A; KMT2F; MGC55143; mKIAA0339; mNSC1; non-selective cation channel 1; Nsccn1; SET domain-containing protein 1A
Gene Aliases: BC010250; KMT2F; mKIAA0339; mNSC1; Nsccn1; Setd1a
UniProt ID: (Mouse) E9PYH6
Entrez Gene ID: (Mouse) 233904
Molecular Function: DNA binding protein methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.