Novus Biologicals
Manufacturer Code:NBP130888
Catalog # NBP130888
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human SET isoform 2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2PP2A HLA-DR-associated protein II I2PP2A I-2PP2A IGAAD Inhibitor of granzyme A-activated DNase inhibitor-2 of protein phosphatase-2A IPP2A2 PHAPIITAF-I Phosphatase 2A inhibitor I2PP2A protein phosphatase type 2A inhibitor protein SET SET nuclear oncogene SET translocation (myeloid leukemia-associated) TAF-IBETA Template-activating factor I Template-Activating Factor-I chromatin remodelling factor; HLA-DR-associated protein II; I-2PP2A; IGAAD; Inhibitor of granzyme A-activated DNase; inhibitor-2 of protein phosphatase-2A; PHAPII; Phosphatase 2A inhibitor I2PP2A; protein phosphatase type 2A inhibitor; Protein SET; SET nuclear oncogene; SET translocation (myeloid leukemia-associated); TAF-I; Template-activating factor I; Template-Activating Factor-I, chromatin remodelling factor
Gene Aliases: 2PP2A; I2PP2A; IGAAD; IPP2A2; PHAPII; SET; TAF-I; TAF-IBETA
UniProt ID: (Human) Q01105
Entrez Gene ID: (Human) 6418
Molecular Function: enzyme modulator phosphatase inhibitor phosphatase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.