Novus Biologicals
Manufacturer Code:NBP144994
Catalog # NBP144994
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to unique epitope on SESN1. The SESN1 peptide was selected from N-terminal amino acids 1-50. This peptide was covalently modified post-synthetically to achieve desired antigenicity. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC138241 MGC142129 p53-regulated protein PA26 PA26p53 activated gene 26 SEST1p53 regulated PA26 nuclear protein sestrin 1 sestrin-1; p53 activated gene 26; p53 regulated PA26 nuclear protein; p53-regulated protein PA26; Sestrin-1
Gene Aliases: PA26; SESN1; SEST1
UniProt ID: (Human) Q9Y6P5
Entrez Gene ID: (Human) 27244
Molecular Function:
oxidoreductase
peroxidase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.