Proteintech
Manufacturer Code:550101AP150UL
Catalog # 50-173-5928
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: p53 activated gene 26; p53 regulated PA26 nuclear protein; p53 regulated protein PA26 PA26 RP11 787I22.1 SESN1 SEST1 sestrin 1; p53-regulated protein PA26; Sestrin-1
Gene Aliases: 1110002G11Rik; AU044290; PA26; SESN1; SEST1
UniProt ID: (Human) Q9Y6P5, (Mouse) P58006
Entrez Gene ID: (Human) 27244, (Mouse) 140742, (Rat) 294518
Molecular Function:
oxidoreductase
peroxidase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.