Invitrogen
Manufacturer Code:MA537842
Catalog # PIMA537842
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0681 |
Immunogen | A synthesized peptide derived from human Hsp47 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 47 kDa heat shock protein; 47 kDa heat shock protein arsenic-transactivated protein 3 AsTP 3 AsTP3 BERF-1 beta enolase repressor factor 1 CBP 1 CBP 2 CBP1 CBP2 cell proliferation-inducing gene 14 protein collagen binding protein 1 collagen binding protein 2 collagen-binding protein Colligen Colligin Colligin 1 Colligin 2 colligin-1 colligin-2 gp46 HGNC 1547 Hsp 47 Hsp47 J6 OI10 PIG 14 PIG14 PPROM RA A47 RA-A47 rheumatoid arthritis antigen A-47 rheumatoid arthritis-related antigen RA-A47 serine (or cysteine) peptidase inhibitor clade H member 1 serine (or cysteine) proteinase inhibitor clade H (heat shock protein 47) member 1 serine (or cysteine) proteinase inhibitor clade H (heat shock protein 47) member 1 (collagen binding protein 1) serine (or cysteine) proteinase inhibitor clade H (heat shock protein 47) member 2 serine (or cysteine) proteinase inhibitor clade H (heat shock protein 47) member 2 (collagen-binding protein 2) serine (or cysteine) proteinase inhibitor clade H member 1 serine protease inhibitor J6 serine proteinase inhibitor clade H (heat shock protein 47) member 1 serpin family H member 1 serpin H1 serpin peptidase inhibitor clade H (heat shock protein 47) member 1 (collagen binding protein 1) serpin peptidase inhibitor clade H member 1 Serpinh1 SERPINH2; Arsenic-transactivated protein 3; AsTP3; beta enolase repressor factor 1; Cell proliferation-inducing gene 14 protein; collagen binding protein 1; Collagen-binding protein; Colligin; colligin-1; colligin-2; rheumatoid arthritis antigen A-47; Rheumatoid arthritis-related antigen RA-A47; serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1; serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1); serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 2; serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 2, (collagen-binding protein 2); Serine protease inhibitor J6; Serpin H1; serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1)
Gene Aliases: AsTP3; BERF-1; CBP1; CBP2; gp46; HSP47; J6; OI10; PIG14; PPROM; RA-A47; SERPINH1; SERPINH2
UniProt ID: (Human) B3KVJ3, (Mouse) P19324
Entrez Gene ID: (Human) 871, (Mouse) 12406
Molecular Function:
enzyme modulator
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.