Abnova
Manufacturer Code:MAB12294
Catalog # 89-934-923
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 134CT3.1.1 |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to amino acids 222-251 at internal region of rat Serpinh1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against synthetic peptide of rat Serpinh1.
For Research Use Only
Protein Aliases: 47 kDa heat shock protein; Cbp2; collagen binding protein 2; Collagen-binding protein; Colligin; GP46; serine (or cysteine) peptidase inhibitor, clade H, member 1; serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1; serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 2; serine (or cysteine) proteinase inhibitor, clade H, member 1; serine proteinase inhibitor, clade H (heat shock protein 47), member 1; Serpin H1; serpin peptidase inhibitor, clade H, member 1; Serpinh2
Gene Aliases: Cbp1; Cbp2; Hsp47; Serpinh1; Serpinh2
UniProt ID: (Rat) P29457
Entrez Gene ID: (Rat) 29345
Molecular Function: enzyme modulator protease inhibitor serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.