Invitrogen
Manufacturer Code:PA595231
Catalog # PIPA595231
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human Hsp47 recombit protein (Position: D247-L418). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 47 kDa heat shock protein; Arsenic-transactivated protein 3; AsTP3; Cell proliferation-inducing gene 14 protein; collagen binding protein 1; collagen binding protein 2; Collagen-binding protein; Colligin; colligin-1; colligin-2; GP46; rheumatoid arthritis antigen A-47; Rheumatoid arthritis-related antigen RA-A47; serine (or cysteine) peptidase inhibitor, clade H, member 1; serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1; serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1); serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 2; serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 2, (collagen-binding protein 2); serine (or cysteine) proteinase inhibitor, clade H, member 1; serine proteinase inhibitor, clade H (heat shock protein 47), member 1; Serpin H1; Serpin H1 47 kDa heat shock protein Arsenic-transactivated protein 3 AsTP3 Cell proliferation-inducing gene 14 protein Collagen-binding protein Colligin Rheumatoid arthritis-related antigen RA-A47 SERPINH1 CBP1 CBP2 HSP47 SERPINH2 PIG14; serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1); serpin peptidase inhibitor, clade H, member 1
Gene Aliases: AsTP3; CBP1; CBP2; gp46; HSP47; OI10; PIG14; PPROM; RA-A47; SERPINH1; SERPINH2
UniProt ID: (Human) B3KVJ3, (Rat) P29457
Entrez Gene ID: (Human) 871, (Rat) 29345
Molecular Function: enzyme modulator protease inhibitor serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.