Invitrogen
Manufacturer Code:PA576659
Catalog # PIPA576659
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human SERPINH1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 47 kDa heat shock protein; Arsenic-transactivated protein 3; AsTP 3AsTP3CBP 1CBP 2CBP1CBP2Cell proliferation-inducing gene 14 proteinCollagen binding protein 1Collagen binding protein 2Collagen binding proteinCollagen-binding protein 2Collagen-binding proteinColligenColligin 1Colligin 2Colligincolligin-1colligin-2gp46Heat shock protein 47HGNC 1547Hsp 47J6OI10PIG 14PIG14PPROMProliferation inducing gene 14Proliferation inducing gene 14 proteinRA A47RA-A47Rheumatoid arthritis antigen A 47rheumatoid arthritis antigen A-47Rheumatoid arthritis related antigen RA A47Serine or cysteine proteinase inhibitor clade H member 1Serine or cysteine proteinase inhibitor clade H member 2SERPH&lowbarHUMANSerpin H1Serpin peptidase inhibitor clade H member 1serpin peptidase inhibitor clade H (heat shock protein 47) member 1 (collagen binding protein 1)Serpin peptidase inhibitor clade H member 1SERPINH1SERPINH2; AsTP3; beta enolase repressor factor 1; Cell proliferation-inducing gene 14 protein; collagen binding protein 1; Collagen-binding protein; Colligin; colligin-1; colligin-2; rheumatoid arthritis antigen A-47; Rheumatoid arthritis-related antigen RA-A47; serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1; serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1); serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 2; serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 2, (collagen-binding protein 2); Serine protease inhibitor J6; Serpin H1; serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1)
Gene Aliases: AsTP3; BERF-1; CBP1; CBP2; gp46; HSP47; J6; OI10; PIG14; PPROM; RA-A47; SERPINH1; SERPINH2
UniProt ID: (Human) B3KVJ3, (Mouse) P19324
Entrez Gene ID: (Human) 871, (Mouse) 12406
Molecular Function:
enzyme modulator
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.