Invitrogen
Manufacturer Code:PA571456
Catalog # PIPA571456
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C terminal region of human SERPINH1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 47 kDa heat shock protein; Arsenic-transactivated protein 3; AsTP3; AsTP3 CBP1 CBP2 HSP47 OI10 PPROM RA-A47 SERPINH2 gp46 47 kDa heat shock protein arsenic-transactivated protein 3 cell proliferation-inducing gene 14 protein colligin-1 colligin-2 rheumatoid arthritis antigen A-47 rheumatoid arthritis-related antigen RA-A47 serine (or cysteine) proteinase inhibitor clade H (; beta enolase repressor factor 1; Cell proliferation-inducing gene 14 protein; collagen binding protein 1; collagen binding protein 2; Collagen-binding protein; Colligin; colligin-1; colligin-2; GP46; heat shock protein 47; HSP47; rheumatoid arthritis antigen A-47; Rheumatoid arthritis-related antigen RA-A47; serine (or cysteine) peptidase inhibitor, clade H, member 1; serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1; serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1); serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 2; serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 2, (collagen-binding protein 2); serine (or cysteine) proteinase inhibitor, clade H, member 1; Serine protease inhibitor J6; serine proteinase inhibitor, clade H (heat shock protein 47), member 1; Serpin H1; serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1); serpin peptidase inhibitor, clade H, member 1
Gene Aliases: AsTP3; BERF-1; CBP1; CBP2; gp46; HSP47; J6; OI10; PIG14; PPROM; RA-A47; SERPINH1; SERPINH2
UniProt ID: (Human) B3KVJ3, (Rat) P29457, (Mouse) P19324
Entrez Gene ID: (Dog) 485187, (Human) 871, (Horse) 100064560, (Pig) 396773, (Rat) 29345, (Mouse) 12406
Molecular Function:
enzyme modulator
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.