OriGene
Manufacturer Code:TA504611
Catalog # 50-167-5263
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2C9 |
Immunogen | Full length human recombit protein of human SERPINE2 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: GDN; GDN GDNPF PI-7 PI7 PN-1 PN1 PNI; Glia-derived nexin; glial-derived neurite promoting factor; Peptidase inhibitor 7; PI-7; PN-1; Protease nexin 1; Protease nexin I; Serpin E2; serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 2
Gene Aliases: GDN; GDNPF; PI-7; PI7; PN-1; PN1; PNI; SERPINE2
UniProt ID: (Human) P07093
Entrez Gene ID: (Human) 5270
Molecular Function:
enzyme modulator
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.