Invitrogen
Manufacturer Code:PA575531
Catalog # PIPA575531
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 157-202 of Human SERPINB7. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Megsin; mesangium predominant gene, megsin; serine (or cysteine) peptidase inhibitor, clade B, member 7; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 7; serine (or cysteine) proteinase inhibitor, clade B, member 7; Serpin B7; Serpin B7 Serpin-B7 Megsin TP55; serpin peptidase inhibitor, clade B (ovalbumin), member 7; TP55
Gene Aliases: 4631416M05Rik; MEGSIN; ovalbumin; PPKN; SERPINB7; TP55
UniProt ID: (Human) O75635, (Mouse) Q9D695
Entrez Gene ID: (Human) 8710, (Rat) 117092, (Mouse) 116872
Molecular Function:
enzyme modulator
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.