Invitrogen
Manufacturer Code:PA572538
Catalog # PIPA572538
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 108-137 amino acids from the Central region of human SERPINB7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Megsin; MEGSIN TP55 mesangium predomit gene megsin serine (or cysteine) proteinase inhibitor clade B (ovalbumin) member 7 serpin B7; mesangium predominant gene, megsin; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 7; Serpin B7; serpin peptidase inhibitor, clade B (ovalbumin), member 7; TP55
Gene Aliases: MEGSIN; PPKN; SERPINB7; TP55
UniProt ID: (Human) O75635
Entrez Gene ID: (Human) 8710
Molecular Function: enzyme modulator protease inhibitor serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.