Abnova
Manufacturer Code:H00008710K
Catalog # 89-203-554
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SERPINB7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human SERPINB7 peptide using ARM Technology.
For Research Use Only
Protein Aliases: DKFZp686D06190 MEGSIN MGC120014 MGC120015; Megsin; mesangium predominant gene, megsin; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 7; Serpin B7; serpin peptidase inhibitor, clade B (ovalbumin), member 7; TP55
Gene Aliases: MEGSIN; PPKN; SERPINB7; TP55
UniProt ID: (Human) O75635
Entrez Gene ID: (Human) 8710
Molecular Function: enzyme modulator protease inhibitor serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.