OriGene
Manufacturer Code:CF504127
Catalog # 50-166-2653
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C5 |
Immunogen | Full length human recombit protein of human SERPINB6 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CAP; CAP DFNB91 MSTP057 PI-6 PI6 PTI SPI3; Cytoplasmic antiproteinase; Peptidase inhibitor 6; PI-6; Placental thrombin inhibitor; protease inhibitor 6 (placental thrombin inhibitor); Proteinase inhibitor 6; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 6; serine (or cysteine) proteinase inhibitor, clade B, member 6a; serine protease inhibitor 3; Serpin B6; serpin peptidase inhibitor, clade B (ovalbumin), member 6
Gene Aliases: 4930482L21Rik; CAP; D330015H01Rik; DFNB91; MSTP057; ovalbumin; PI-6; PI6; PTI; SERPINB6; Serpinb6a; SPI3
UniProt ID: (Human) P35237, (Mouse) Q60854
Entrez Gene ID: (Human) 5269, (Mouse) 20719
Molecular Function:
enzyme modulator
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.