OriGene
Manufacturer Code:CF800076
Catalog # 50-166-4838
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4B3 |
Immunogen | Human recombit protein fragment corresponding to amino acids 77-306 of human SERPINB1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EI; EI ELANH2 HEL-S-27 HEL57 LEI M/NEI MNEI PI-2 PI2; epididymis luminal protein 57; epididymis secretory protein Li 27; LEI; Leukocyte elastase inhibitor; Leukocyte elastase inhibitor A; Monocyte/neutrophil elastase inhibitor; Peptidase inhibitor 2; PI-2; protease inhibitor 2 (anti-elastase), monocyte/neutrophil derived; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 1; Serine protease inhibitor EIA; Serpin B1; Serpin B1a; serpin peptidase inhibitor, clade B (ovalbumin), member 1
Gene Aliases: EI; ELANH2; HEL-S-27; HEL57; LEI; M/NEI; MNEI; PI-2; PI2; SERPINB1; Serpinb1a
UniProt ID: (Human) P30740, (Rat) Q4G075
Entrez Gene ID: (Human) 1992, (Dog) 488191, (Rat) 291091
Molecular Function:
enzyme modulator
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.