Invitrogen
Manufacturer Code:PA521992
Catalog # PIPA521992
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 371 of TBG (Uniprot IDNo.P05543) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RP1-82J11.2 TBG T4-binding globulin serine (or cysteine) proteinase inhibitor clade A (alpha-1 antiproteinase antitrypsin) member 7 serpin A7 thyroxin-binding globulin thyroxine-binding globulin; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 7; Serpin A7; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 7; T4-binding globulin; thyroxin-binding globulin; Thyroxine-binding globulin
Gene Aliases: SERPINA7; TBG
UniProt ID: (Human) P05543
Entrez Gene ID: (Human) 6906
Molecular Function: enzyme modulator protease inhibitor serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.