Invitrogen
Manufacturer Code:PA518954
Catalog # PIPA518954
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (QSPETPAPQNQTSR) corresponding to the N-terminus amino acids of SERPINA10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein Z-dependent protease inhibitor; PZ-dependent protease inhibitor; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10; Serpin A10; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10; SERPINA10 ZPI serpin peptidase inhibitor clade A (alpha-1 antiproteinase antitrypsin) member 10 PZI protein Z-dependent protease inhibitor protein Z-dependent protease inhibitorprecursor serine (or cysteine) proteinase inhibitor clade A (alpha-1 antiproteinase antitrypsin) member 10
Gene Aliases: PZI; SERPINA10; UNQ707/PRO1358; ZPI
UniProt ID: (Human) Q9UK55
Entrez Gene ID: (Human) 51156
Molecular Function: enzyme modulator protease inhibitor serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.