Invitrogen
Manufacturer Code:PA5113515
Catalog # PIPA5113515
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Protein Z-dependent protease inhibitor protein (165-444AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: plasma protein associated with liver regeneration Protein Z-dependent protease inhibitor PZ-dependent protease inhibitor PZI Rasp1 RASP-1 Regeneration-associated serpin 1 serine (or cysteine) peptidase inhibitor clade A (alpha-1 antiproteinase antitrypsin) member 10 serine (or cysteine) proteinase inhibitor clade A (alpha-1 antiproteinase antitrypsin) member 10 Serpin A10 serpin family A member 10 serpin peptidase inhibitor clade A (alpha-1 antiproteinase antitrypsin) member 10 serpin peptidase inhibitor clade A member 10 Serpina10 UNQ707/PRO1358 ZPI; Protein Z-dependent protease inhibitor; PZ-dependent protease inhibitor; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10; Serpin A10; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10
Gene Aliases: PZI; SERPINA10; UNQ707/PRO1358; ZPI
UniProt ID: (Human) Q9UK55, (Mouse) Q8R121
Entrez Gene ID: (Human) 51156, (Mouse) 217847
Molecular Function: enzyme modulator protease inhibitor serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.