Invitrogen
Manufacturer Code:PA558615
Catalog # PIPA558615
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SERP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RAMP4 ribosome associated membrane protein 4 ribosome-attached membrane protein 4; ribosome associated membrane protein 4; Ribosome-attached membrane protein 4; Stress-associated endoplasmic reticulum protein 1
Gene Aliases: D3Ucla1; RAMP4; SERP1
UniProt ID: (Human) Q9Y6X1, (Rat) Q9R2C1, (Mouse) Q9Z1W5
Entrez Gene ID: (Human) 27230, (Rat) 80881, (Mouse) 28146
Molecular Function: RNA binding protein chaperone nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.