Novus Biologicals
Manufacturer Code:NBP19859220UL
Catalog # NBP19859220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this antibody is SERP1 - N-terminal region. Peptide sequence IRMANEKHSKNITQRGNVAKTSRNAPEEKASVGPWLLALFIFVVCGSAIF. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ43424 MGC117327 MGC133321 RAMP4MGC133322 ribosome associated membrane protein 4 Ribosome-attached membrane protein 4 stress-associated endoplasmic reticulum protein 1; ribosome associated membrane protein 4; Ribosome-attached membrane protein 4; Stress-associated endoplasmic reticulum protein 1
Gene Aliases: RAMP4; SERP1
UniProt ID: (Human) Q9Y6X1
Entrez Gene ID: (Human) 27230
Molecular Function: RNA binding protein chaperone nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.