Novus Biologicals
Manufacturer Code:NBP297673V
Catalog # NB046341
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human SERINC3 fragment. |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AIGP1 DIFF33tumor differentially expressed 1 placental transmembrane protein (mouse testicular tumor differentiallyexpressed) SBBI99 serine incorporator 3 TDE TDE1Tumor differentially expressed protein 1 TMS-1 transmembrane protein SBBI99 tumour differentially expressed 1; Serine incorporator 3; Tumor differentially expressed protein 1
Gene Aliases: AIGP1; DIFF33; SBBI99; SERINC3; TDE; TDE1; TMS-1
UniProt ID: (Human) Q13530
Entrez Gene ID: (Human) 10955
Molecular Function:
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.