Proteintech
Manufacturer Code:202671AP150UL
Catalog # 50-173-2805
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AIGP-1; AIGP1 DIFF33 SBBI99 SERINC3 serine incorporator 3 TDE TDE1 TMS 1; Axotomy-induced glyco/Golgi protein 1; Axotomy-induced glycoprotein 1; Membrane protein TMS-1; Serine incorporator 3; tumor differentially expressed 1; Tumor differentially expressed protein 1
Gene Aliases: AA959945; AA960047; AIGP1; DIFF33; SBBI99; SERINC3; TDE; TDE1; TMS-1; Tms1
UniProt ID: (Human) Q13530, (Mouse) Q9QZI9
Entrez Gene ID: (Human) 10955, (Mouse) 26943, (Rat) 296350
Molecular Function:
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.