Proteintech
Manufacturer Code:202661AP150UL
Catalog # 50-173-2804
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FKSG84 PRO0899 SERINC2 serine incorporator 2 TDE2 TDE2L UNQ263/PRO300; Serine incorporator 2; tumor differentially expressed protein 2; Tumor differentially expressed protein 2-like
Gene Aliases: 2310004K20Rik; AW121759; FKSG84; PRO0899; SERINC2; TDE2; TDE2L; UNQ263/PRO300
UniProt ID: (Human) Q96SA4, (Mouse) Q8K0E7
Entrez Gene ID: (Human) 347735, (Mouse) 230779, (Rat) 313057
Molecular Function: transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.