Novus Biologicals
Manufacturer Code:NB300622
Catalog # NB300622
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | VE121G9 |
Immunogen | Purified rabbit skeletal muscle sarcoplasmic reticulum. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ATP2A ATPase Ca++ transporting cardiac muscle fast twitch 1 Calcium pump 1 Calcium-transporting ATPase sarcoplasmic reticulum type fast twitch skeletalmuscle isoform EC 3.6.3 Endoplasmic reticulum class 1/2 Ca(2+) ATPase sarcoplasmic/endoplasmic reticulum calcium ATPase 1 SERCA1EC 3.6.3.8 SR Ca(2+)-ATPase 1; ATPase, Ca++ transporting, cardiac muscle, fast twitch 1; Calcium pump 1; Calcium-transporting ATPase sarcoplasmic reticulum type, fast twitch skeletal muscle isoform; Endoplasmic reticulum class 1/2 Ca(2+) ATPase; Sarcoplasmic/endoplasmic reticulum calcium ATPase 1; SERCA1; SR Ca(2+)-ATPase 1
Gene Aliases: ATP2A; ATP2A1; SERCA1
UniProt ID: (Human) O14983
Entrez Gene ID: (Human) 487
Molecular Function:
cation transporter
hydrolase
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.