Invitrogen
Manufacturer Code:PA545141
Catalog # PIPA545141
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SERAC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein SERAC1; Protein SERAC1-like protein; serine active site containing 1; Serine active site-containing protein 1
Gene Aliases: 4930511N22Rik; D17Ertd141e; RGD1565064; SERAC1
UniProt ID: (Bovine) Q2TBM9, (Human) Q96JX3, (Mouse) Q3U213
Entrez Gene ID: (Bovine) 519860, (Dog) 476255, (Rabbit) 100338574, (Horse) 100051068, (Pig) 100523935, (Human) 84947, (Mouse) 321007, (Guinea pig) 100716991, (Rat) 499015
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.