Proteintech
Manufacturer Code:153331AP150UL
Catalog # 50-173-0155
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SEPX1 Fusion Protein Ag7391 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HSPC270 MSRB1 Selenoprotein X selenoprotein X 1 SELR SELX SEPX1; Methionine-R-sulfoxide reductase B1; MsrB1; Selenoprotein R; Selenoprotein X; selenoprotein X 1; selenoprotein X, 1; SelR; SelX
Gene Aliases: D17Wsu82e; HSPC270; MSRB1; SELR; SELX; SepR; SEPX1
UniProt ID: (Human) Q9NZV6, (Rat) Q52KJ8, (Mouse) Q9JLC3
Entrez Gene ID: (Human) 51734, (Rat) 685059, (Mouse) 27361
Molecular Function:
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.