Proteintech
Manufacturer Code:665421IG150UL
Catalog # 50-173-6962
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1G12D1 |
Immunogen | SEPT7 Fusion Protein Ag4309 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CDC10 (cell division cycle 10, S. cerevisiae, homolog); CDC10 (cell division cycle 10, S.cerevisiae, homolog); CDC10 CDC10 protein homolog CDC3 Nbla02942 SEPT7 SEPT7A septin 7; CDC10 protein homolog; cell division cycle 10 homolog; septin 7 variant 4; Septin-7
Gene Aliases: CDC10; CDC3; E430034N22; NBLA02942; SEPT7; SEPT7A; SEPTIN7
UniProt ID: (Human) Q16181, (Mouse) O55131, (Rat) Q9WVC0
Entrez Gene ID: (Human) 989, (Mouse) 235072, (Rat) 64551
Molecular Function: G-protein cytoskeletal protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.