Abnova
Manufacturer Code:PAB18783
Catalog # 89-120-340
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 14 amino acids at C-terminus of human SEPT4 . |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SEPT4 .
For Research Use Only
Protein Aliases: Apoptosis-related protein in the TGF-beta signaling pathway; ARTS; BRADEION; Bradeion beta; Brain protein H5; CE5B3; CE5B3 beta; Cell division control-related protein 2; Cerebral protein 7; H5; hCDCREL-2; hucep-7; MART; Peanut-like protein 2; PNUTL2; SEP4; Septin-4; septin-M
Gene Aliases: ARTS; BRADEION; C17orf47; CE5B3; H5; hCDCREL-2; hucep-7; MART; PNUTL2; SEP4; SEPT4; SEPTIN4
UniProt ID: (Human) O43236
Entrez Gene ID: (Human) 5414
Molecular Function:
G-protein
cytoskeletal protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.