Invitrogen
Manufacturer Code:PA569493
Catalog # PIPA569493
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse SEPN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Selenoprotein N; Selenoprotein N 1 FLJ24021 MDRS1 RSMD1 RSS SELN CFTD; selenoprotein N, 1; SelN
Gene Aliases: 1110019I12Rik; AI414492; CFTD; MDRS1; RSMD1; RSS; SELENON; SELN; SEPN1
UniProt ID: (Human) Q9NZV5, (Mouse) D3Z2R5
Entrez Gene ID: (Bovine) 616379, (Human) 57190, (Mouse) 74777, (Rat) 362624
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.