Invitrogen
Manufacturer Code:PA581257
Catalog # PIPA581257
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to the N-terminus of the Human SENP8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: deneddylase 1; Deneddylase-1; NEDD8 specific-protease cysteine 2; NEDD8-specific protease 1; Protease, cysteine 2; Sentrin-specific protease 8; Sentrin-specific protease 8 Sentrin/SUMO-specific protease SENP8 Protease cysteine 2 NEDD8-specific protease 1 deneddylase-1 DEN1 NEDP1 PRSC2; Sentrin/SUMO-specific protease SENP8; SUMO sentrin specific protease family member 8; SUMO/sentrin specific peptidase family member 8
Gene Aliases: DEN1; FKSG8; NEDP1; PRSC2; SENP8
UniProt ID: (Human) Q96LD8
Entrez Gene ID: (Human) 123228
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.