Invitrogen
Manufacturer Code:PA569704
Catalog # PIPA569704
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human SENP6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2810017C20Rik; SENP6 SUSP1 SUMO1/sentrin specific peptidase 6 RP1-134M13.1 FLJ11355 FLJ11887 KIAA0389 KIAA0797 SSP1 2810017C20Rik SUMO-1-specific protease SUMO1/sentrin specific protease 6; Sentrin-specific protease 6; Sentrin-specific protease 6-like protein; Sentrin/SUMO-specific protease SENP6; SUMO-1-specific protease 1; SUMO/sentrin specific peptidase 6; SUMO/sentrin specific protease 6; SUMO1/sentrin specific peptidase 6; SUMO1/sentrin specific protease 6; Sumo1/sentrin/SMT3 specific peptidase 6
Gene Aliases: 2810017C20Rik; E130319N12Rik; FKSG6; KIAA0797; mKIAA0797; SENP6; SSP1; SUSP1
UniProt ID: (Human) Q9GZR1, (Mouse) Q8BUL4
Entrez Gene ID: (Human) 26054, (Dog) 481883, (Pig) 100155462, (Bovine) 533853, (Horse) 100068828, (Rabbit) 100353104, (Rat) 300860, (Mouse) 215351
Molecular Function:
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.