Abnova
Manufacturer Code:PAB7602
Catalog # 89-133-407
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human SENP6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of SENP6.
For Research Use Only
Protein Aliases: 2810017C20Rik; FLJ11355; FLJ11887; KIAA0389; KIAA0797; Sentrin-specific protease 6; Sentrin/SUMO-specific protease SENP6; SSP1; SUMO-1-specific protease 1; SUMO1/sentrin specific protease 6; SUSP1
Gene Aliases: FKSG6; KIAA0797; SENP6; SSP1; SUSP1
UniProt ID: (Human) Q9GZR1
Entrez Gene ID: (Human) 26054
Molecular Function:
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.