Novus Biologicals
Manufacturer Code:NBP156590
Catalog # NBP156590
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SENP3(SUMO1/sentrin/SMT3 specific peptidase 3) The peptide sequence was selected from the N terminal of SENP3. Peptide sequence PPPKPRLKSGGGFGPDPGSGTTVPARRLPVPRPSFDASASEEEEEEEEEE. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZP586K0919 DKFZp762A152 EC 3.4.22 EC 3.4.22.- sentrin/SUMO-specific protease 3 Sentrin/SUMO-specific protease SENP3 sentrin-specific protease 3 SMT3IP1 SSP3DKFZp586K0919 SUMO1/sentrin/SMT3 specific peptidase 3 SUMO1/sentrin/SMT3 specific protease 3 SUMO-1-specific protease 3 SUSP3; Sentrin-specific protease 3; sentrin/SUMO-specific protease 3; Sentrin/SUMO-specific protease SENP3; SUMO-1-specific protease 3
Gene Aliases: SENP3; SMT3IP1; SSP3; SUSP3; Ulp1
UniProt ID: (Human) Q9H4L4
Entrez Gene ID: (Human) 26168
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.