Invitrogen
Manufacturer Code:PA572915
Catalog # PIPA572915
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Gel purified rat p47 seminal vesicle protein I (secreted Semenogelin I) from rodent seminal vesicle secretion fluid was used as immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-inhibin-31; Alpha-inhibin-92; Cancer/testis antigen 103; CT103 SEMG SGI dJ172H20.2 cancer/testis antigen 103 semen coagulating protein semenogelin-1; semen coagulating protein; semenoclotin; Semenogelin I; Semenogelin-1; Seminal basic protein; seminal vesicle protein, secretion 1; seminal vesicle protein, secretion 2; seminal vesicle secretory protein 2; seminal vesicle secretory protein II; SGI; SgI-29; SVS II
Gene Aliases: BB115391; CT103; dJ172H20.2; RATSVPIIA; SEMG; SEMG1; SGI; Svp1; SVPIIA; Svs2; SVS2P; Svs2p2; SvsII
UniProt ID: (Human) P04279
Entrez Gene ID: (Human) 6406, (Rat) 25013, (Mouse) 53878
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.