Invitrogen
Manufacturer Code:PA5111590
Catalog # PIPA5111590
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human SEMG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-inhibin-31; Alpha-Inhibin-31 Alpha-Inhibin-92 BB115391 Cancer/testis antigen 103 CT103 dJ172H20.2 MGC14719 RATSVPIIA Semenogelin I Semenogelin-1 SEMG SEMG1 Seminal basic protein SGI Svp1 Svp-1 Svp5 SVPIIA Svs2 SVS2P Svs2p2 Svs5 SvsII; Alpha-inhibin-92; Cancer/testis antigen 103; semen coagulating protein; Semenogelin I; Semenogelin-1; Seminal basic protein; SGI; SgI-29
Gene Aliases: CT103; dJ172H20.2; SEMG; SEMG1; SGI
UniProt ID: (Human) P04279
Entrez Gene ID: (Human) 6406
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.