Invitrogen
Manufacturer Code:PA543381
Catalog # PIPA543381
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SEMA6D |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Sema domain Transmembrane domain (TM) and cytoplasmic domain (semaphorin) 6D FLJ11598 KIAA1479; sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6D; Semaphorin-6D
Gene Aliases: 1110067B02Rik; AA409156; D330011G23; KIAA1479; mKIAA1479; SEMA6D; wu:fi06a12; wu:fi76a10; zgc:56170
UniProt ID: (Human) Q8NFY4, (Mouse) Q76KF0
Entrez Gene ID: (Horse) 100055875, (Dog) 100855988, (Bovine) 518458, (Rabbit) 100353114, (Human) 80031, (Guinea pig) 100715046, (Rat) 311384, (Mouse) 214968, (Zebrafish) 406272
Molecular Function:
membrane-bound signaling molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.