Invitrogen
Manufacturer Code:PA547791
Catalog # PIPA547791
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHO-derived recombit human Semaphorin 5A Glu23-Thr765 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: M-Sema D; sema domain, seven thrombospondin repeats (type 1 and type 1-like), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 5A; Sema F; sema5a {ECO:0000312|Ensembl:ENSRNOP00000016506}; SEMAF semF sema F semaphorin F semaphorin-5A semaphorin-F; semaphorin 5A; semaphorin F; Semaphorin-5A; semaphorin-5A {ECO:0000250|UniProtKB:Q13591}; Semaphorin-F
Gene Aliases: 5930434A13; 9130201M22Rik; AI464145; SEMA5A; SEMAF; semF
UniProt ID: (Human) Q13591, (Rat) D3ZTD8
Entrez Gene ID: (Human) 9037, (Mouse) 20356, (Rat) 310207
Molecular Function:
membrane-bound signaling molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.