Invitrogen
Manufacturer Code:PA5116598
Catalog # PIPA5116598
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 330-500 of human SEMA5A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5930434A13 9130201M22Rik AI464145 Ensembl:ENSRNOP00000016506} M-Sema D sema domain seven thrombospondin repeats (type 1 and type 1-like) transmembrane domain (TM) and short cytoplasmic domain (semaphorin) 5A Sema F SEMA5A sema5a {ECO:0000312 SEMAF semaphorin 5A semaphorin F semaphorin-5A semaphorin-5A {ECO:0000250 Semaphorin-F semF UniProtKB:Q13591}; M-Sema D; sema domain, seven thrombospondin repeats (type 1 and type 1-like), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 5A; Sema F; semaphorin F; Semaphorin-5A; Semaphorin-F
Gene Aliases: 5930434A13; 9130201M22Rik; AI464145; SEMA5A; SEMAF; semF
UniProt ID: (Human) Q13591
Entrez Gene ID: (Human) 9037, (Mouse) 20356
Molecular Function: membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.