Abnova
Manufacturer Code:MAB20878
Catalog # 89-337-375
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | SEMA4D/933 |
Immunogen | Recombinant protein corresponding to human SEMA4D |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
Mouse Monoclonal Antibody.
Protein Aliases: A8; BB18; C9orf164 CD100 FLJ33485 FLJ34282 FLJ39737 FLJ46484 M-sema-G MGC169138 MGC169141 SEMAJ coll-4; CD100; GR3; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4D; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, 4D; Semaphorin-4D
Gene Aliases: C9orf164; CD100; coll-4; M-sema-G; SEMA4D; SEMAJ
UniProt ID: (Human) Q92854
Entrez Gene ID: (Human) 10507
Molecular Function:
membrane-bound signaling molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.