OriGene
Manufacturer Code:TA809851
Catalog # 50-168-3054
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI8G5 |
Immunogen | Full length human recombit protein of human SEMA4D produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: A8; BB18; C9orf164 CD100 COLL4 M-sema-G SEMAJ coll-4; CD100; GR3; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4D; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, 4D; Semaphorin-4D
Gene Aliases: C9orf164; CD100; coll-4; M-sema-G; SEMA4D; SEMAJ
UniProt ID: (Human) Q92854
Entrez Gene ID: (Human) 10507
Molecular Function:
membrane-bound signaling molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.