Invitrogen
Manufacturer Code:PA552788
Catalog # PIPA552788
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SEMA4C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: M-Sema F; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4C; SEMA4C SEMACL1 Semaf SEMAI FLJ20369 KIAA1739 MGC126382 MGC126383 M-Sema F M-SEMA-F sema domain immunoglobulin domain (Ig) transmembrane domain (TM) and shortcytoplasmic domain (semaphorin) 4C SEMACL1 Semacl1 SEMAF Semaf SEMAI semaphorin-4C; Semaphorin-4C
Gene Aliases: KIAA1739; M-SEMA-F; SEMA4C; SEMACL1; SEMAF; SEMAI; UNQ5855/PRO34487
UniProt ID: (Human) Q9C0C4
Entrez Gene ID: (Human) 54910
Molecular Function:
membrane-bound signaling molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.