Abnova
Manufacturer Code:PAB20534
Catalog # 89-122-039
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human SEMA4C. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant SEMA4C.
For Research Use Only
Protein Aliases: FLJ20369; KIAA1739; M-Sema F; M-SEMA-F; MGC126382; MGC126383; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4C; SEMACL1; SEMAF; SEMAI; Semaphorin-4C
Gene Aliases: KIAA1739; M-SEMA-F; SEMA4C; SEMACL1; SEMAF; SEMAI; UNQ5855/PRO34487
UniProt ID: (Human) Q9C0C4
Entrez Gene ID: (Human) 54910
Molecular Function: membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.