Invitrogen
Manufacturer Code:PA588896
Catalog # PIPA588896
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 300-400 of human SEMA3A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: collapsin-1; Sema D; sema domain, immunoglobulin domain (Ig), short basic domain, sec; Sema III; Semaphorin 3a; Semaphorin III; Semaphorin-3A; Semaphorin-D
Gene Aliases: coll-1; Hsema-I; SEMA1; Sema3a; Semad; SemD
UniProt ID: (Mouse) O08665, (Rat) Q63548
Entrez Gene ID: (Mouse) 20346, (Rat) 29751
Molecular Function: membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.