Abnova
Manufacturer Code:PAB7888
Catalog # 89-133-469
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal region of human SEMA3A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SEMA3A.
For Research Use Only
Protein Aliases: coll-1; collapsin 1; Hsema-I; Hsema-III; MGC133243; sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3A; Sema III; SEMA1; SEMAD; SEMAIII; SEMAL; semaphorin D; Semaphorin III; semaphorin L; Semaphorin-3A; SemD
Gene Aliases: coll-1; COLL1; HH16; Hsema-I; Hsema-III; SEMA1; SEMA3A; SEMAD; SEMAIII; SEMAL; SemD
UniProt ID: (Human) Q14563
Entrez Gene ID: (Human) 10371
Molecular Function: membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.