Invitrogen
Manufacturer Code:PA552686
Catalog # PIPA552686
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SELS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: histocompatibility 47; Minor histocompatibility antigen H47; Selenoprotein S; Selenoprotein S AD-015 ADO15 MGC104346 MGC2553 SBBI8 SEPS1 VIMP SELS; SelS; Sg2; Tanis; valosin-containing protein-interacting membrane protein; VCP-interacting membrane protein; VCP-interacting membrane selenoprotein
Gene Aliases: 1500011E07Rik; AD-015; ADO15; C78786; H-4; H-47; H4; H47; SBBI8; SELENOS; SELS; SEPS1; sg2; VIMP
UniProt ID: (Human) Q9BQE4, (Rat) Q8VHV8, (Mouse) Q9BCZ4
Entrez Gene ID: (Human) 55829, (Rat) 286900, (Mouse) 109815
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.