Invitrogen
Manufacturer Code:PA543131
Catalog # PIPA543131
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SELS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chondroitin sulfate synthase 1; histocompatibility 47; Minor histocompatibility antigen H47; Selenoprotein S; Selenoprotein S AD-015 ADO15 MGC104346 MGC2553 SBBI8 SEPS1 VIMP SELS; SelS; Sg2; Tanis; valosin-containing protein-interacting membrane protein; VCP interacting membrane selenoprotein; VCP-interacting membrane protein; VCP-interacting membrane selenoprotein
Gene Aliases: 1500011E07Rik; AD-015; ADO15; C78786; CHSY1; H-4; H-47; H4; H47; SBBI8; SELENOS; SELS; SEPS1; sg2; VIMP; wu:fc16a03; zgc:136970
UniProt ID: (Bovine) Q2KI76, (Human) Q9BQE4, (Rat) Q8VHV8, (Mouse) Q9BCZ4, (Zebrafish) Q0VFV6
Entrez Gene ID: (Dog) 607791, (Horse) 100057595, (Goat) 102179048, (Rabbit) 100347427, (Bovine) 512276, (Guinea pig) 100713682, (Human) 55829, (Rat) 286900, (Mouse) 109815, (Zebrafish) 735249
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.